missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF625 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543607
This item is not returnable.
View return policy
Description
Peptide sequence: VGHASLNRHH RADTGHKPYE YQEYGQKPYK CTYCKKAFSD LPYFRTHEWA Sequence homology: Horse: 85%; Human: 100%.
ZNF625 may be involved in transcriptional regulation.
Specifications
| ZNF625 | |
| Polyclonal | |
| Unconjugated | |
| ZNF625 | |
| Zinc finger protein 625; ZNF625 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 90589 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96I27 | |
| ZNF625 | |
| Synthetic peptide directed towards the middle region of human ZNF625. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction