missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF625 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112807
This item is not returnable.
View return policy
Description
ZNF625 Polyclonal Antibody for Western Blot, ELISA
ZNF625 may be involved in transcriptional regulation.
Specifications
| ZNF625 | |
| Polyclonal | |
| Unconjugated | |
| ZNF625 | |
| Zinc finger protein 625; ZNF625 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 90589 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96I27 | |
| ZNF625 | |
| Recombinant Human Zinc finger protein 625 protein (1-306AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction