missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Aldolase C Protein

Product Code. 18604148 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.50mg
This item is not returnable. View return policy

Product Code. 18604148

Brand: Novus Biologicals™ NBP249693

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 20% glycerol, 2 mM DTT, 0.1 M NaCl
Gene ID (Entrez) 230
Molecular Weight (g/mol) MolecularWeight-theroretical: 39.4 kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Gene Alias ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C
Gene Symbol ALDOC
Research Category Core ESC Like Genes, Stem Cell Markers
Purity or Quality Grade >90%, by SDS-PAGE
Protein Aldolase C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.