missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF629 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 624.00
Specifications
| Antigen | ZNF629 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18223245
|
Novus Biologicals
NBP2-58789 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604147
|
Novus Biologicals
NBP2-58789-25ul |
25 μL |
N/A
|
N/A | |||||
Description
ZNF629 Polyclonal specifically detects ZNF629 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ZNF629 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 23361 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MEPETALWGPDLQGPEQSPNDAHRGAESENEEESPRQESSGEEIIMGDPAQSPESKDSTEMSLERSSQD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| DNA-binding protein, KIAA0326, zinc finger protein 629, zinc finger protein 65, ZNF, ZNF65 | |
| ZNF629 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title