missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF638 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584342
This item is not returnable.
View return policy
Description
Immunogen sequence: EPIKSVNQSI NQTVSQTMSQ SLIPPSMNQQ PFSSELISSV SQQERIPHEP VINSSNVHVG SRGSKKNYQS QADIPIRSPF GIVKASWLPK F Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 68%.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. NP220 (Nuclear protein 220), also designated Zinc finger protein 638 (ZNF638) or Cutaneous T-cell lymphoma-associated antigen se33-1, is a 1978 amino acid protein that contains one matrin-type zinc finger and two RRM (RNA recognition motif) domains, suggesting a role in transcriptional regulation. NP220 binds to double-stranded DNA fragments by recognizing clusters of cytidines. NP220 interacts with FHL-2 and is also thought to be phosphorylated by ATM or ATR upon DNA damage. It exists as five isoforms as a result of alternative splicing events. Isoform five of NP220 is a tumor-associated antigen found in several cutaneous T-cell lymphoma (CTCL), and in particular in mycosis fungoides patients and in Sezary syndrome patients.
Specifications
| ZNF638 | |
| Polyclonal | |
| Unconjugated | |
| ZNF638 | |
| AI323587; CTCL tumor antigen se33-1; CTCL-associated antigen se33-1; Cutaneous T-cell lymphoma-associated antigen se33-1; mNP220; Np220; NP220 nuclear protein; nuclear protein 220; Zfml; Zfp638; Zinc finger matrin-like protein; zinc finger protein 638; zinc finger, matrin-like; Znf638 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14966 | |
| ZNF638 | |
| Recombinant protein corresponding to Human ZNF638. Recombinant protein control fragment (Product #RP-106774). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction