missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF672 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15178
This item is not returnable.
View return policy
Beschreibung
ZNF672 Polyclonal specifically detects ZNF672 in Mouse samples. It is validated for Western Blot.
Spezifikation
| ZNF672 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| FLJ22301, zinc finger protein 672 | |
| Rabbit | |
| 50 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot | |
| 0.93mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q499Z4 | |
| ZNF672 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF672. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79894 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur