Learn More
ZNF687 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41128
 View more versions of this product
Product Code. 18000434
  • ONLINE EXCLUSIVE PRICE
    € 546.00
    € 515.55 / 0.10mg
    Save € 30.45
    5% Off
Description
ZNF687 Polyclonal specifically detects ZNF687 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF687
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
ZNF687
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZNF687. The immunogen is located within the first 50 amino acids of ZNF687. Amino Acid Squence: HSGPEENEGPGGPGKPE
0.1 mg
Primary
ZNF687 antibody is human specific. At least three isoforms of ZNF687 are known to exist; this antibody will detect the the two largest isoforms.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1 mg/mL
ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
KIAA1441DKFZp781I1719, zinc finger protein 687
Rabbit
Affinity Purified
RUO
57592