Learn More
ZNF687 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41175JF646
Product Code. 18746422
  • ONLINE EXCLUSIVE PRICE
    € 630.00
    € 595.35 / 0.10mL
    Save € 34.65
    5% Off
Description
ZNF687 Polyclonal antibody specifically detects ZNF687 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF687
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
57592
Store at 4°C in the dark.
IgG
Western Blot, ELISA, Immunofluorescence
Janelia Fluor 646
KIAA1441DKFZp781I1719, zinc finger protein 687
Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZNF687. The immunogen is located within amino acids 1160 - 1210 of ZNF687. Amino Acid Squence: DGEEEAPPSRSDPDGGD
0.1 mL