Learn More
ZNF69 Monoclonal Antibody (OTI3C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808649S
This item is not returnable.
View return policy
Description
ZNF69 Monoclonal antibody specifically detects ZNF69 in Human samples. It is validated for Western BlotSpecifications
| ZNF69 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UC07 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7620 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3C6 | |
| Unconjugated | |
| ZNF69 | |
| Cos5; hZNF3; zinc finger protein 69; ZNF3; ZNF69 | |
| Full length human recombinant protein of human ZNF69 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |