missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF70 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110608
This item is not returnable.
View return policy
Description
Immunogen sequence: MEVPPATKFG ETFAFENRLE SQQGLFPGED LGDPFLQERG LEQMAVIYKE IPLGEQDEEN DDYEGNFSLC SSPVQHQSIP PGTRPQDDEL FGQTFLQKSD LSMCQIIHSE EPSPCDCAET DRGDSGPNAP HRTPQPAKPY.
The function of the ZNF70 gene has not yet been determined.
Specifications
| ZNF70 | |
| Polyclonal | |
| Unconjugated | |
| ZNF70 | |
| Cos17; Zinc finger protein 70; zinc finger protein N27C7-1; ZNF70 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7621 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.36 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9UC06 | |
| ZNF70 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human ZNF70 (NP_068735.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction