missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF703 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32138F
This item is not returnable.
View return policy
Description
ZNF703 Polyclonal antibody specifically detects ZNF703 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ZNF703 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| FLJ14299, ZEPPO1, Zinc finger elbow-related proline domain protein 1, zinc finger protein 703, ZNF503L, ZPO1 | |
| The immunogen this antibody was made to, maps to a region between residue 100 to 150 of human Zinc Finger Protein 703 using the numbering given in entry NP_079345.1 (GeneID 80139). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 80139 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction