missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF703 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112813
This item is not returnable.
View return policy
Description
ZNF703 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
ZNF703 belongs to the Elbow/Noc family. ZNF703 contains 1 C2H2-type zinc finger. ZNF703 may function as a transcriptional repressor.
Specifications
| ZNF703 | |
| Polyclonal | |
| Unconjugated | |
| Znf703 | |
| 1110032O19Rik; AI430822; AL022941; cb514; Csmn1; End2; ES neuronal differentiation 2; fc45d06; fk28e09; NH11; nlz; nlz1; nocA related zinc finger 1; nocA-like zinc finger protein 1; nocA-like Zn-finger protein; nocA-related zinc finger protein 1; noz1; si:dkey-158j20.1; wu:fc45d06; wu:fk28e09; Zeppo1; Zfp703; zgc:111927; Zinc finger elbow-related proline domain protein 1; zinc finger protein 703; zinc finger protein Noz1; ZNF503L; Znf703; Zpo1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 680717, 80139 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.65 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H7S9 | |
| Zfp703, Znf703 | |
| Recombinant Human Zinc finger protein 703 protein (218-286AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction