missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF711 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118374
This item is not returnable.
View return policy
Description
ZNF711 Polyclonal Antibody for Western Blot, ICC/IF, IP
This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
Specifications
| ZNF711 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9Y462 | |
| ZNF711 | |
| E. coli-derived Human ZNF711 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| ZNF711 | |
| 2810409C01Rik; A230078I01Rik; CMPX1; dJ75N13.1; dJ75N13.1 (znf6-like); MRX97; Zfp711; Zinc finger protein 6; zinc finger protein 6 (CMPX1); zinc finger protein 711; ZNF4; ZNF5; ZNF6; ZNF711 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7552 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction