missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF728 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145173

Product Code. 18066071

  • € 604.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HELVKEPPGR TGHELWLRKL ELSLGTAIGT KVCRPASIAL NGYHSPGLWK TQDLSQTVAG RSLG Antigen sequence identity: Mouse 31%; Rat 27%.

ZNF728 is a zinc finger protein that is encoded by the ZNF728 gene in humans. It contains multiple zinc finger domains and is involved in transcriptional regulation and gene expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF728
Polyclonal
Unconjugated
ZNF728
Zinc finger protein 728
Rabbit
Affinity Chromatography
RUO
388523
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
IgG
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P0DKX0
ZNF728
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Recombinant protein control fragment (Product #RP-109939).
100 μL
Primary
Human
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.