missing translation for 'onlineSavingsMsg'
Learn More

ZNF729 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15653465
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15653465 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15653465 Supplier Invitrogen Supplier No. PA549169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ZNF729 Polyclonal Antibody

ZNF729 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF729
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF729
Gene Accession No. A6NN14
Gene Alias Zinc finger protein 729; ZNF729
Gene Symbols ZNF729
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 112-139 amino acids from the N-terminal region of human ZNF729.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100287226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.