missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF75 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88035
This item is not returnable.
View return policy
Description
ZNF75 Polyclonal specifically detects ZNF75 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZNF75 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| D8C6, MGC126327, Zinc finger protein 75, zinc finger protein 75 (D8C6), zinc finger protein 75D, Zinc finger protein 82, ZNF75ZNF82 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7626 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZNF75D | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ETVISLGLKLKNDTGNDHPISVSTSEIQTSGCEVSKKTRMKIAQKTMGRENPGDTHSVQKWHRAFPRKKRKKPATCKQELPKLMDLHGKGPTGEKP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only
Spot an opportunity for improvement?