missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF75D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538622
This item is not returnable.
View return policy
Description
Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 75 (ZNF75), also known as ZNF82, is a 289 amino acid member of the Krueppel C2H2-type zinc finger protein family. Localized to the nucleus, ZNF75 contains five C2H2- type zinc fingers and one KRAB domain through which it is thought to be involved in DNA-binding and transcriptional regulation.
Specifications
| ZNF75D | |
| Polyclonal | |
| Unconjugated | |
| ZNF75D | |
| Zinc finger protein 75, Zinc finger protein 75D, Zinc finger protein 82, Zinc finger protein 82, ZN75D, ZNF75D, ZNF82, | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7626 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51815 | |
| ZNF75D | |
| A synthetic peptide derived from the internal region of human ZNF75 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction