missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF763 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21922-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF763 may be involved in transcriptional regulation.Specifications
| ZNF763 | |
| Polyclonal | |
| Unconjugated | |
| ZNF763 | |
| zinc finger protein 763, ZNF, ZNF440L, ZNF763 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 284390 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q0D2J5 | |
| ZNF763 | |
| ZNF763 Fusion Protein Ag16440 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction