missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SPEGYLRGNM SENEEEEISQ QEGSGDYEVE EIPFGLEPQS PGFEPQSPEF EPQSPRFEPE SPGFESRSPG LVPPSPEFAP RS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
May be involved in transcriptional regulation.
Specifications
Specifications
| Antigen | ZNF768 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF768 |
| Gene Accession No. | Q9H5H4 |
| Gene Alias | Zinc finger protein 768; ZNF768 |
| Gene Symbols | ZNF768 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
