missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF780A Polyclonal Antibody

ZNF780A Polyclonal Antibody
Brand: Invitrogen PA524037
This item is not returnable.
View return policy
Description
ZNF780A Polyclonal Antibody for Western Blot, Flow
Specifications
| ZNF780A | |
| Polyclonal | |
| Unconjugated | |
| ZNF780A | |
| Zinc finger protein 780A; ZNF780; ZNF780A | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 284323 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O75290 | |
| ZNF780A | |
| KLH conjugated synthetic peptide between 54-83 amino acids from the N-terminal region of human ZNF780A. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction