missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF783 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00155060-B01P
This item is not returnable.
View return policy
Description
ZNF783 Polyclonal antibody specifically detects ZNF783 in Human, Rat samples. It is validated for Western Blot
Specifications
| ZNF783 | |
| Polyclonal | |
| Western Blot 1:500 | |
| DKFZp667J212, DKFZp762P2111, FLJ12914, FLJ16713, FLJ36716, LOC155060 AI894139 pseudogene | |
| ZNF783 (AAI31570.1, 1 a.a. - 281 a.a.) full-length human protein. MAEAAPARDPETDKHTEDQSPSTPLPQPAAEKNSYLYSTEITLWTVVAAIQALEKKVDSCLTRLLTLEGRTGTAEKKLADCEKTAVEFGNQLEGKWAVLGTLLQEYGLLQRRLENVENLLRNRNFWILRLPPGSKGEAPKVPVTFDDVAVYFSELEWGKLEDWQKELYKHVMRGNYETLVSLDYAISKPDILTRIERGEEPCLDRWGQEKGNEVEVGRPRMMGTGLPPYPEHLTSPLSPAQEELKEGQAPKQQQDSEARVAPAGPEAGLALRTDLQGEAQI | |
| 0.05 mg | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 155060 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction