missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF787 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78198PE
This item is not returnable.
View return policy
Description
ZNF787 Polyclonal antibody specifically detects ZNF787 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ZNF787 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| TIP20, transcription termination factor I interacting peptide 20, TTF-I interacting peptide 20, TTF-I-interacting peptide 20, zinc finger protein 787 | |
| The immunogen for this product maps to a region between residue 200 and 250 of human Zinc Finger Protein 787 using the numbering given in entry NP_001002836.2 (GeneID 126208). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 126208 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction