missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF787 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567581
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
May be involved in transcriptional regulation.
Specifications
| ZNF787 | |
| Polyclonal | |
| Unconjugated | |
| ZNF787 | |
| 2210018M03Rik; TIP20; transcription termination factor I interacting peptide 20; TTF-I-interacting peptide 20; Zfp787; Zinc finger protein 787; ZNF787 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 126208, 67109 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6DD87, Q8BIF9 | |
| Zfp787, ZNF787 | |
| Synthesized peptide derived from human ZNF787. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction