missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF804B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15773301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15773301 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15773301 Supplier Invitrogen™ Supplier No. PA555383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KEVNISPSHL ESVLHNTISI NSKILQDKHD SIDETLEDSI GIHASFSKSN IHLSDVDFTP TSREKETRNT LKNTLENCVN HPCQANASFS PPNIYNHSDA RISECLDEFS SLEP Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 53%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF804B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF804B
Gene Accession No. A4D1E1
Gene Alias zinc finger 804B; zinc finger protein 804B; ZNF804B
Gene Symbols ZNF804B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF804B. Recombinant protein control fragment (Product #RP-92821).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219578
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.