missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF81 Antibody (PCRP-ZNF81-2G2), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14088MFV450
This item is not returnable.
View return policy
Description
ZNF81 Monoclonal antibody specifically detects ZNF81 in Human samples. It is validated for Flow Cytometry
Specifications
| ZNF81 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 347344 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| PCRP-ZNF81-2G2 | |
| Flow Cytometry | |
| dJ54B20.6, FLJ44367, HFZ20X-linked 45, zinc finger protein 81, zinc finger protein 81 (HFZ20) | |
| Recombinant full-length human ZNF81 protein (Uniprot: P51508) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction