missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF853 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA564337

Product Code. 15732102

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MLHQPTPRNR GLTARMEVGP ATETFVLELR CLEDGGPGPD TLSGGSGGSE SQ Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 75%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF853
Polyclonal
Unconjugated
ZNF853
Zinc finger protein 853; ZNF853
Rabbit
Antigen affinity chromatography
RUO
54753
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P0CG23
ZNF853
Recombinant protein corresponding to Human ZNF853. Recombinant protein control fragment (Product #RP-104185).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.