missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF9 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566556

Product Code. 15939254

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GMRSRGRGGFT SDRGFQFVSS SLPDICYRCG ESG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

This gene encodes a nucleic-acid binding protein with seven zinc-finger domains. The protein has a preference for binding single stranded DNA and RNA. The protein functions in cap-independent translation of ornithine decarboxylase mRNA, and may also function in sterol-mediated transcriptional regulation. A CCTG expansion in the first intron of this gene results in myotonic dystrophy type 2. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF9
Polyclonal
Unconjugated
CNBP
AA408710; CCHC-type zinc finger nucleic acid binding protein; CCHC-type zinc finger, nucleic acid binding protein; cellular nucleic acid binding protein; cellular nucleic acid binding protein 1; cellular nucleic acid-binding protein; CNBP; CNBP1; DM2; erythroid differentiation-related; HGNC:13164; PROMM; RNF163; sterol regulatory element-binding protein; ZCCHC22; zinc finger protein 273; zinc finger protein 9; zinc finger protein 9 (a cellular retroviral nucleic acid binding protein); Znf9
Rabbit
Antigen Affinity Chromatography
RUO
7555
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P62633
CNBP
Recombinant Human ZNF9. Recombinant protein control fragment (Product #RP-105254).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.