missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF96 Antibody (PCRP-ZSCAN12-1A9), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24347FR
This item is not returnable.
View return policy
Description
ZNF96 Monoclonal antibody specifically detects ZNF96 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF96 | |
| Monoclonal | |
| DyLight 680 | |
| KIAA0426dJ29K1.2, ZFP96, zinc finger and SCAN domain containing 12, Zinc finger protein 305ZNF305, Zinc finger protein 96ZNF96zinc finger and SCAN domain-containing protein 12, ZNF29K1 | |
| Recombinant fragment (around aa37-132) of human ZNF96 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| PCRP-ZSCAN12-1A9 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 9753 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction