missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNFX1, Mouse anti-Human, Clone: 6F9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00057169-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: PGRHPRANNHPAAYWQREERFRAMGRNPHQGRRNQEGHASDEARDQRHDQENDTRWRNGNQDCRNRRPPWSNDNFQQWRTPHQKPTEQPQQAKKLGYKFLESpecifications
| ZNFX1 | |
| Monoclonal | |
| Unconjugated | |
| zinc finger, NFX1-type containing 1 | |
| FLJ39275/MGC131926 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 6F9 | |
| In 1x PBS, pH 7.4 | |
| NM_021035 | |
| ZNFX1 | |
| ZNFX1 (NP_066363.1, 50 a.a. ∼ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 57169 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction