missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNRD1 Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505067S
This item is not returnable.
View return policy
Description
ZNRD1 Monoclonal antibody specifically detects ZNRD1 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| ZNRD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q70YC5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 22891 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| ZNF365 | |
| 1110014N07Rik; DNA-directed RNA polymerase I subunit RPA12; HTEX6; HTEX-6; hZR14; nuclear RNA polymerase I small specific subunit; nuclear RNA polymerase I small specific subunit Rpa12 like; RNA polymerase I small specific subunit Rpa12; Rpa12; Tctex6; tctex-6; TEX6; transcription-associated zinc ribbon protein; zinc ribbon domain containing 1; zinc ribbon domain containing, 1; zinc ribbon domain-containing protein 1; ZNRD1; ZR14 | |
| Human recombinant protein fragment corresponding to amino acids 108-407 of human ZNF365 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction