Learn More
ZNRD1 Mouse anti-Human, Clone: OTI1G2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505067
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a DNA-directed RNA polymerase I subunit. The encoded protein contains two potential zinc-binding motifs and may play a role in regulation of cell proliferation. The encoded protein may be involved in cancer and human immunodeficiency virus progression. Alternative splicing results in multiple transcript variants.Specifications
| ZNRD1 | |
| Monoclonal | |
| Unconjugated | |
| ZNRD1 | |
| HTEX-6, HTEX6, Rpa12, TCTEX6, TEX6, ZR14, hZR14, tctex-6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 30834 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1G2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9P1U0 | |
| ZNRD1 | |
| Full length human recombit protein of human ZNRD1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.