missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNRD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585046
This item is not returnable.
View return policy
Description
Immunogen sequence: GTAMPMSVEE GPECQGPVVD RRCPRCGHEG MAYHTRQMRS ADEGQTVFYT CTNCKFQEKE DS Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 87%.
This gene encodes a DNA-directed RNA polymerase I subunit. The encoded protein contains two potential zinc-binding motifs and may play a role in regulation of cell proliferation. The encoded protein may be involved in cancer and human immunodeficiency virus progression. Alternative splicing results in multiple transcript variants.
Specifications
| ZNRD1 | |
| Polyclonal | |
| Unconjugated | |
| ZNRD1 | |
| 1110014N07Rik; DNA-directed RNA polymerase I subunit RPA12; HTEX6; HTEX-6; hZR14; nuclear RNA polymerase I small specific subunit; nuclear RNA polymerase I small specific subunit Rpa12 like; RNA polymerase I small specific subunit Rpa12; Rpa12; Tctex6; tctex-6; TEX6; transcription-associated zinc ribbon protein; zinc ribbon domain containing 1; zinc ribbon domain containing, 1; zinc ribbon domain-containing protein 1; ZNRD1; ZR14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P1U0 | |
| ZNRD1 | |
| Recombinant protein corresponding to Human ZNRD1. Recombinant protein control fragment (Product #RP-108697). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction