missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNRF2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28697B
This item is not returnable.
View return policy
Description
ZNRF2 Polyclonal antibody specifically detects ZNRF2 in Human samples. It is validated for Western Blot
Specifications
| ZNRF2 | |
| Polyclonal | |
| Western Blot | |
| EC 6.3.2.-, Protein Ells2, RING finger protein 202, RNF202E3 ubiquitin-protein ligase ZNRF2, zinc and ring finger 2, zinc finger/RING finger 2, Zinc/RING finger protein 2 | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human zinc and ring finger 2 using the numbering given in entry NP_667339.1 (GeneID 223082). | |
| 0.1 mL | |
| Zinc Finger | |
| 223082 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction