missing translation for 'onlineSavingsMsg'
Learn More

ZnT-8/SLC30A8 Antibody (815039) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30164670 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30164670 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30164670 Supplier Novus Biologicals Supplier No. FAB7936JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZnT-8/SLC30A8 Monoclonal antibody specifically detects ZnT-8/SLC30A8 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZnT-8/SLC30A8
Applications Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 815039
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias solute carrier family 30 (zinc transporter), member 8, Solute carrier family 30 member 8, zinc transporter 8, zinc transporter ZnT-8, ZNT8ZnT-8
Host Species Mouse
Immunogen E. coli-derived recombinant human ZnT-8, Lys268-Pro359, Accession # Q8IWU4
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 169026
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.