Learn More
Invitrogen™ ZO-3 Monoclonal Antibody (K24021_24G11), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | ZO-3 |
|---|---|
| Clone | K24021_24G11 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30206125
|
Invitrogen™
MA556309 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30206991
|
Invitrogen™
MA555929 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: DSDEDDGPQR VEEVDQGRGY DGDSSSGSGR SWDERSRRPR PGRRGRAGSH GRRSPGGGSE ANGLALVSGF KRLPRQDVQM KPVKSVLVKR RDSEEFGVKL GSQIFIKHIT DSGLAARHRG LQEGDLILQI NGVSSQNLSL NDTRRLIEKS EGKLSLLVLR DRGQFLVNIP PAVSDSDSSP LEDISDLASE LSQAPPSHIP PPPRHAQRSP EASQTDSPVE SPRLRRESSV DSRTISEPDE QRSELPRESS YDIYRVPSSQ SMEDRGYSPD TRVVRFLKGK SIGLRLAGGN DVGIFVSGVQ AGSPADGQGI QEGDQILQVN DVPFQNLTRE EAVQFLLGLP PGEEMELVTQ
ZO-3 Monoclonal antibody specifically detects ZO-3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ZO-3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| tight junction protein 3; tight junction protein 3 (zona occludens 3); tight junction protein ZO-3; TJP3; TJP-3; ZO3; ZO-3; Zona occludens protein 3; zonula occludens protein 3 | |
| Recombinant human TJP3 (immunogen range is 111-460). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TJP3 |
| K24021_24G11 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O95049 | |
| 27134 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.