Learn More
Zofenopril calcium, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Specifications
Specifications
| Chemical Name or Material | Zofenopril calcium |
| CAS | 81938-43-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H22Ca0.5NO4S2 |
| Quantity | 10 mg |
| Synonym | SQ26991 |
| Solubility Information | DMSO : 2 mg/mL (4.46 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(N(C[C@@H](SC1=CC=CC=C1)C2)[C@@H]2C([O-])=O)[C@H](C)CSC(C3=CC=CC=C3)=O.[0.5Ca2+] |
| Molecular Weight (g/mol) | 448.58 |
| Show More |
Research purposes only