Learn More
Zofenopril calcium, MedChemExpress
Product Code. 16458406
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458406 100 mg 100mg
16444437 50 mg 50mg
16428568 10 mg 10mg
16411038 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16458406 Supplier Medchem Express Supplier No. HYB0655
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Specifications
Chemical Name or Material Zofenopril calcium
CAS 81938-43-4
Color White
Purity Grade Notes Research
Molecular Formula C22H22Ca0.5NO4S2
Quantity 100 mg
Synonym SQ26991
Solubility Information DMSO : 2 mg/mL (4.46 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N(C[C@@H](SC1=CC=CC=C1)C2)[C@@H]2C([O-])=O)[C@H](C)CSC(C3=CC=CC=C3)=O.[0.5Ca2+]
Molecular Weight (g/mol) 448.58
Formula Weight 448.58
Percent Purity 99.56%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less
Research purposes only