missing translation for 'onlineSavingsMsg'
Learn More

Zoligratinib, MedChemExpress

Product Code. 16473907
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473907

Brand: Medchem Express HY19957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zoligratinib (Debio 1347) is an orally available and selective FGFR inhibitor with IC50s of 9.3, 7.6, and 22 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.

Zoligratinib (Debio 1347) is an orally available and selective FGFR inhibitor with IC50s of 9.3, 7.6, and 22 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.

Specifications

Chemical Name or Material Zoligratinib
Physical Form Solid
CAS 1265229-25-1
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H16N6O
For Use With (Application) Cancer-Kinase/protease
Synonym Debio 1347 CH5183284
Solubility Information DMSO : 25 mg/mL (70.15 mM; ultrasonic and warming and heat to 50°C)
SMILES O=C(C1=C(N)N(C2=CC=C3N=C(C)NC3=C2)N=C1)C(N4)=CC5=C4C=CC=C5
Molecular Weight (g/mol) 356.38
Formula Weight 356.38
Percent Purity 99.32%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.