Learn More
Zolmitriptan-d6 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | Zolmitriptan-D6 |
| CAS | 1217644-84-2 |
| Molecular Formula | C16H15D6N3O2 |
| Quantity | 1 mg |
| Synonym | (4S)-4-[[3-[2-(Dimethyl-d6-amino)ethyl]-1H-indol-5-yl]methyl-2-oxazolidinone, BW-311C90-d6, Zomig-d6 |
| InChI Formula | InChI=1S/C16H21N3O2/c1-19(2)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13-10-21-16(20)18-13/h3-4,8-9,13,17H,5-7,10H2,1-2H3,(H,18,20)/t13-/m0/s1/i1D3,2D3 |
| SMILES | O=C1OC[C@@H](N1)CC2=CC(C(CCN(C([2H])([2H])[2H])C([2H])([2H])[2H])=CN3)=C3C=C2 |
| IUPAC Name | (4S)-4-[[3-[2-[bis(trideuteriomethyl)amino]ethyl]-1H-indol-5-yl]methyl]-1,3-oxazolidin-2-one |
| Molecular Weight (g/mol) | 293.39 |
| Formula Weight | 293.201 |