missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde)
Specifications
Specifications
| Chemical Name or Material | Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde) |
| CAS | 400777-11-9 |
| Analyte or Component Names | Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H14 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Zolpaldehyde, 6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde |
| InChI Formula | InChI=1S/C16H14N2O/c1-11-3-6-13(7-4-11)16-14(10-19)18-9-12(2)5-8-15(18)17-16/h3-10H,1-2H3 |
| SMILES | Cc1ccc(cc1)c2nc3ccc(C)cn3c2C=O |
| IUPAC Name | 6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carbaldehyde |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction