Learn More
Zolpidem Phenyl-4-carboxylic Acid Ethyl Ester-d6, TRC
Product Code. 30454012
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454012 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30454012 Supplier Toronto Research Chemicals Supplier No. TRCZ6500222.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Zolpidem Phenyl-4-carboxylic Acid Ethyl Ester-d6
CAS 1216455-48-9
Molecular Formula C21H17D6N3O3
Quantity 2.5 mg
Synonym 4-[3-[2-(Dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoic Acid Ethyl Ester-d6.
InChI Formula InChI=1S/C21H23N3O3/c1-5-27-21(26)16-9-7-15(8-10-16)20-17(12-19(25)23(3)4)24-13-14(2)6-11-18(24)22-20/h6-11,13H,5,12H2,1-4H3/i3D3,4D3
SMILES [2H]C([2H])([2H])N(C(=O)Cc1c(nc2ccc(C)cn12)c3ccc(cc3)C(=O)OCC)C([2H])([2H])[2H]
IUPAC Name ethyl 4-[3-[2-[bis(trideuteriomethyl)amino]-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoate
Molecular Weight (g/mol) 371.47
Formula Weight 371.2116