missing translation for 'onlineSavingsMsg'
Learn More

Zomepirac Sodium Salt, TRC

Product Code. 30451186
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451186 Supplier Toronto Research Chemicals Supplier No. TRCZ675000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zomepirac Sodium
CAS 64092-48-4
Molecular Formula C15 H13 Cl N O3 . Na
Quantity 100 mg
Synonym Zomepirac Sodium, 1H-Pyrrole-2-acetic acid','5-(4-chlorobenzoyl)-1,4-dimethyl-','sodium salt (9CI), McN 2783-21-98, Sodium zomepirac, Zomax, Zomepirac sodium, Zomepirac sodium salt
InChI Formula InChI=1S/C15H14ClNO3.Na/c1-9-7-12(8-13(18)19)17(2)14(9)15(20)10-3-5-11(16)6-4-10;/h3-7H,8H2,1-2H3,(H,18,19);/q;+1/p-1
SMILES [Na+].Cc1cc(CC(=O)[O-])n(C)c1C(=O)c2ccc(Cl)cc2
IUPAC Name sodium;2-[5-(4-chlorobenzoyl)-1,4-dimethylpyrrol-2-yl]acetate
Molecular Weight (g/mol) 313.71
Formula Weight 313.0482
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.