missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZONAB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535151
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat based on sequence homology.
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.
Specifications
| ZONAB | |
| Polyclonal | |
| Unconjugated | |
| YBX3 | |
| cold shock domain protein A; cold shock domain-containing protein A; cold-shock domain containing A1; cold-shock domain protein A; Csda; CSDA1; Dbpa; DNA-binding protein A; Dpba; MSY3; MSY4; Muscle Y-box protein YB2; oxyR; RYB-A; single-strand DNA-binding protein NF-GMB; Y box binding protein 3; Y box protein 3; Yb2; Y-box binding protein 3; Y-box protein 3; Y-box-binding protein 3; Y-box-binding protein A; Ybx3; ZO-1-associated nucleic acid-binding protein; ZONAB; ZONAB A; ZONAB B | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 56449, 8531 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P16989, Q9JKB3 | |
| YBX3 | |
| KLH conjugated synthetic peptide between 152-181 amino acids from the central region of human CSDA. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction