Learn More
Buerkle™ Zone Sampler Spiralus
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
770 mL
1100 mL
Length (Metric):
145 cm
200 cm
Unit Size:
Each
Description
- Representative sample of the container layers.
- aluminum, anodised
- Tip PA
- Low weight
Specifications
Specifications
| Product Type | Zone Sampler Spiralus |
| Material | Aluminum |
| Includes | 8 openings |
| Capacity (English) | 26 oz. |
| Capacity (Metric) | 770 mL |
| For Use With (Application) | Sampling Coarse-Grained Bulk Goods, Seeds and Pellets at Locations Such as Soil Heaps, Warehouses, Trucks, Railway Goods Wagons |
| Diameter (Metric) Outer | 38 mm |
| Length (Metric) | 145 cm |