missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZPK Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA597067

Product Code. 16539901

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MACLHETRTP SPSFGGFVST LSEASMRKLD PDTSDCTPEK DLTPTQCVLR DVVPLGGQGG GGPSPSPGGE PPPEPFANSV LQLHEQDAGG PGGAAGSPES RASRVRADEV RLQCQSGSGF LEGLFGCLRP VWTMIGKAYS TEHKQQQEDL WEVPFEEILD LQWVGSGAQG AVFLGRFHGE EVAVKKVRDL KETDIKHLRK LKHPNIITFK; Positive Samples: SH-SY5Y, HT-1080; Cellular Location: Cytoplasm, Membrane, Peripheral membrane protein.

ZPK is a member of serine/threonine protein kinase family. This kinase contains a leucine-zipper domain, and is predominately expressed in neuronal cells. The phosphorylation state of this kinase in synaptic terminals was shown to be regulated by membrane depolarization via calcineurin. This kinase forms heterodimers with leucine zipper containing transcription factors, such as cAMP responsive element binding protein (CREB) and MYC, and thus may play a regulatory role in PKA or retinoic acid induced neuronal differentiation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZPK
Polyclonal
Unconjugated
MAP3K12
DLK; Dual leucine zipper bearing kinase; dual leucine zipper kinase; dual leucine zipper kinase DLK; leucine zipper protein kinase; leucine-zipper protein kinase; M3KC; MAP3K12; MAPK-upstream kinase; MEKK12; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); mitogen activated protein kinase kinase kinase 12; mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; PK; protein kinase MUK; zipper (leucine) protein kinase; Zpk; ZPKP1
Rabbit
Affinity chromatography
RUO
7786
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q12852
MAP3K12
Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human MAP3K12 (NP_0011804401).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.