missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan ZPure LS PolyGas II Purifier 1/4 SS Compression Fittings
Brand: Trajan 202288L-SS
ZPure LS PolyGas II Purifier 1/4 SS Compression Fittings
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ZPure LS PolyGas II Purifier 1/4 SS Compression Fi ttings
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Carbon, Quartz
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction