missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZRANB2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55323R
This item is not returnable.
View return policy
Description
ZRANB2 Polyclonal antibody specifically detects ZRANB2 in Human samples. It is validated for Western Blot
Specifications
| ZRANB2 | |
| Polyclonal | |
| Western Blot | |
| Oocyte-specific maternal effect factor, zygote arrest 1, zygote arrest protein 1 | |
| The immunogen recognized by this antibody maps to a region between residue 270 and the C-terminus (residue 320) of human zinc-finger protein 265 (zinc-finger, splicing) using the numbering given in entry NP_005446.2 (GeneID 9406). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9406 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction