missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZRANB3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93301-0.02ml
This item is not returnable.
View return policy
Description
ZRANB3 Polyclonal antibody specifically detects ZRANB3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZRANB3 | |
| Polyclonal | |
| Western Blot 1:1000-1:3000 | |
| MGC105033, MGC75012,4933425L19Rik, zinc finger Ran-binding domain-containing protein 3, zinc finger, RAN-binding domain containing 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 818-1077 of human ZRANB3 (NP_001273497.1). ITKQQTKQNCTKRYITKEDVAVASMDKVKNVGGHVRLITKESRPRDPFTKKLLEDGACVPFLNPYTVQADLTVKPSTSKGYLQAVDNEGNPLCLRCQQPTCQTKQACKANSWDSRFCSLKCQEEFWIRSNNSYLRAKVFETEHGVCQLCNVNAQELFLRLRDAPKSQRKNLLYATWTSKLPLEQLNEMIRNPGEGHFWQVDHIKPVYGGGGQCSLDNLQTLCTVCHKERTARQAKERSQVRRQSLASKHGSDITRFLVKK | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84083 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction