missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZRSR2 Monoclonal Antibody (2H5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520357
This item is not returnable.
View return policy
Description
Peptide Sequence: GRQLQCEFCP VTRWKMAICG LFEIQQCPRG KHCNFLHVFR NPNNEFWEAN RDIYLSPDRT GSSFGKNSER RERMGHHDDY YSRLRGRRNP SPDHSYKRNG.
This gene encodes an essential splicing factor. The encoded protein associates with the U2 auxiliary factor heterodimer, which is required for the recognition of a functional 3' splice site in pre-mRNA splicing, and may play a role in network interactions during spliceosome assembly.
Specifications
| ZRSR2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q15696 | |
| ZRSR2 | |
| ZRSR2 (NP_005080.1, 294 a.a. ~ 393 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 2H5 | |
| Unconjugated | |
| ZRSR2 | |
| 35kDa; 5031411E02Rik; A230052C13Rik; C77286; CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2; Renal carcinoma antigen NY-REN-20; RGD1559763; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF), 35 kDa, related sequence 2; U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2; U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2; U2 small nuclear RNA auxiliary factor 1-like 2; U2(RNU2) small nuclear RNA auxiliary factor 1-like 2; U2AF1L2; U2AF1RS2; U2AF1-RS2; U2AF35-related protein; URP; ZC3H22; zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2; zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2; zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2; ZRSR2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8233 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction