missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZRSR2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15317664
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15317664 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15317664 Supplier Invitrogen™ Supplier No. PA535248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZRSR2 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes an essential splicing factor. The encoded protein associates with the U2 auxiliary factor heterodimer, which is required for the recognition of a functional 3' splice site in pre-mRNA splicing, and may play a role in network interactions during spliceosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZRSR2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZRSR2
Gene Accession No. Q15696
Gene Alias 35kDa; 5031411E02Rik; A230052C13Rik; C77286; CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2; Renal carcinoma antigen NY-REN-20; RGD1559763; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF), 35 kDa, related sequence 2; U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2; U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2; U2 small nuclear RNA auxiliary factor 1-like 2; U2(RNU2) small nuclear RNA auxiliary factor 1-like 2; U2AF1L2; U2AF1RS2; U2AF1-RS2; U2AF35-related protein; URP; ZC3H22; zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2; zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2; zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2; ZRSR2
Gene Symbols ZRSR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 453-482 amino acids from the C-terminal region of human ZRSR2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.